This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00996923_m1
See other NEURL1B GE Assays ›
Gene Symbol:
NEURL1B
Gene Name:
neuralized E3 ubiquitin protein ligase 1B
Gene Aliases:
NEURL3, hNeur2, neur2
Chromosome Location:
Chr.5: 172641266 - 172691540 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00996923_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NEURL1B
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142651.2 NP_001136123.1 2 - 3 722 78
GenBank mRNA GQ414757.1 - 2 - 3 722 78

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0500009455.hg.1
This assay targets the same location as HTA transcript cluster(s) TC05000948.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ubiquitin-protein ligase

Gene Ontology Categories:

Function(s) Process(es)

Notch signaling pathway
protein ubiquitination
ubiquitin-dependent endocytosis
protein binding
zinc ion binding
ligase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR05013815.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013815.hg.1 ENST00000520919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013815.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05013815.hg.1 TC05000948.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR05013814.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013814.hg.1 ENST00000520919.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013814.hg.1 ENST00000522853.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013814.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05013814.hg.1 TC05000948.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05013816.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013816.hg.1 ENST00000522853.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR05013816.hg.1 NM_001142651.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR05013816.hg.1 TC05000948.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay TC0500009455.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166225.hg.1 TC0500009455.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166228.hg.1 TC0500009455.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 ENST00000522853.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 NM_001308177.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166224.hg.1 TC0500009455.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 ENST00000369800.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 ENST00000520919.5 Ensembl
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 NM_001142651.2 Refseq
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 NM_001308178.1 Refseq
Affymetrix Human Clariom D Assay PSR0500166227.hg.1 TC0500009455.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline