| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013818.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013818.hg.1 |
ENST00000520919.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013818.hg.1 |
ENST00000522853.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013818.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013818.hg.1 |
TC05000948.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013819.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013819.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05013819.hg.1 |
TC05000948.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166230.hg.1 |
TC0500009455.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000520919.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
ENST00000522853.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC0500009455.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
ENST00000369800.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
NM_001142651.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
NM_001308177.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
NM_001308178.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500166231.hg.1 |
TC0500009455.hg.1 |
Clariom D TC |