This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00997068_g1
See other IL32 GE Assays ›
Gene Symbol:
IL32
Gene Name:
interleukin 32
Gene Aliases:
IL-32alpha, IL-32beta, IL-32delta, IL-32gamma, NK4, TAIF, TAIFa, TAIFb, TAIFc, TAIFd
Chromosome Location:
Chr.16: 3065312 - 3069819 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00997068_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: IL32
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001012631.1 NP_001012649.1 2 - 3 246 76
NM_001012634.1 NP_001012652.1 1 - 2 46 76
GenBank mRNA AK296143.1 - 1 - 2 51 76
AK310053.1 - 1 - 2 45 76
BM822755.1 - 2 - 3 154 76

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600006658.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000099.hg.1

Additional Information:

This assay is in the 5'UTR region of the AK296143.1,NM_001012631.1,NM_001012634.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene IL32

Gene Ontology Categories:

Function(s) Process(es)

defense response
immune response
cell adhesion
cytokine activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000008180.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000325568.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000382213.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000396887.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000396890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000440815.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000444393.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000525643.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000526464.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000528163.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000529550.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000529699.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000530538.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000530890.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000533097.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000534507.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000548476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000548652.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000549213.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000551122.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000551513.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000552356.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000552664.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 ENST00000552936.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012631.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012632.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012633.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012634.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012635.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012636.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_001012718.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 NM_004221.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001782.hg.1 TC16000099.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000008180.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000396890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000444393.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000525643.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000526464.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000528163.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000529550.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000530538.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000548476.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000548652.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000549213.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 ENST00000552936.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 NM_001012631.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 NM_001012632.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 NM_001012634.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16001799.hg.1 TC16000099.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000008180.13 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000325568.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000382213.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000396887.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000396890.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000440815.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000444393.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000525228.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000525377.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000525643.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000526464.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000528163.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000529550.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000529699.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000530538.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000530890.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000532086.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000532247.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000533097.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000534507.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000534748.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000548476.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000548652.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000548807.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000549213.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000551122.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000551513.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000552356.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000552664.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000552936.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000613483.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012631.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012632.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012633.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012634.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012635.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012636.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012718.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001308078.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_004221.4 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000008180.13 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000325568.9 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000382213.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000396887.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000396890.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000440815.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000444393.7 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000525228.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000525377.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000525643.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000526464.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000528163.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000529550.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000529699.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000530538.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000530890.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000532086.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000532247.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000533097.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000534507.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000534748.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000548476.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000548652.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000548807.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000549213.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000551122.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000551513.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000552356.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000552664.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000552936.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 ENST00000613483.4 Ensembl
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012631.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012632.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012633.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012634.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012635.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012636.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001012718.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_001308078.1 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 NM_004221.4 Refseq
Affymetrix Human Clariom D Assay PSR1600147418.hg.1 TC1600006658.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0