This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00997541_m1
See other MAGT1 GE Assays ›
Gene Symbol:
MAGT1
Gene Name:
magnesium transporter 1
Gene Aliases:
IAP, MRX95, OST3B, PRO0756, XMEN, bA217H1.1
Chromosome Location:
Chr.X: 77826364 - 77895568 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00997541_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MAGT1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_032121.5 NP_115497.4 6 - 7 913 67
GenBank mRNA AK027632.1 - 4 - 5 430 67
AK075394.1 - 6 - 7 776 67
AK223131.1 - 6 - 7 776 67
AK294943.1 - 6 - 7 938 67
AK314309.1 - 6 - 7 776 67
AL136636.1 - 6 - 7 784 67
AY358691.1 - 6 - 7 763 67
BC041014.1 - 6 - 7 760 3834858 67
BC060842.1 - 6 - 7 776 30338916 67

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00010130.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001174.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene MAGT1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

protein N-linked glycosylation
magnesium ion transport
protein N-linked glycosylation via asparagine
cognition
transmembrane transport
magnesium ion transmembrane transport
magnesium ion transmembrane transporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X015816.hg.1 TC0X001174.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X015816.hg.1 ENST00000358075.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015816.hg.1 ENST00000453109.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X015816.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay PSR0X00169172.hg.1 TC0X00010130.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000358075.10 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000373336.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000476168.1 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000610432.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 ENST00000618282.4 Ensembl
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq
Affymetrix Human Clariom D Assay TC0X00010130.hg.1 NM_032121.5 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0