This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00998266_m1
See other EXTL1 GE Assays ›
Gene Symbol:
EXTL1
Gene Name:
exostosin like glycosyltransferase 1
Gene Aliases:
EXTL
Chromosome Location:
Chr.1: 26021749 - 26036464 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00998266_m1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EXTL1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004455.2 NP_004446.2 9 - 10 2552 96
XM_005245779.3 XP_005245836.1 8 - 9 2475 96
GenBank mRNA BC065528.1 - 9 - 10 2398 6150218 96
U67191.1 - 9 - 10 2551 96

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100007434.hg.1
This assay targets the same location as HTA transcript cluster(s) TC01000332.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EXTL1

Panther Classification:

Molecular Function -

glycosyltransferase

Gene Ontology Categories:

Function(s) Process(es)

skeletal system development
glycosaminoglycan biosynthetic process
protein glycosylation
heparan sulfate proteoglycan biosynthetic process
glucuronosyl-N-acetylglucosaminyl-proteoglycan 4-alpha-N-acetylglucosaminyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR01005079.hg.1 TC01000332.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR01005079.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR01005079.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000481377.5 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 ENST00000484339.1 Ensembl
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay TC0100007434.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 ENST00000374280.3 Ensembl
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 ENST00000470037.1 Ensembl
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 NM_004455.2 Refseq
Affymetrix Human Clariom D Assay PSR0100151835.hg.1 TC0100007434.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0