This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00999523_m1
See other CEACAM18 GE Assays ›
Gene Symbol:
CEACAM18
Gene Name:
carcinoembryonic antigen related cell adhesion molecule 18
Gene Aliases:
-
Chromosome Location:
Chr.19: 51478543 - 51491261 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00999523_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CEACAM18
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001278392.1 NP_001265321.1 2 - 3 398 73
XM_011527258.2 XP_011525560.1 2 - 3 498 73

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900008649.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay TC1900008649.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay TC1900008649.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay PSR1900167557.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900167557.hg.1 NM_001278392.1 Refseq
Affymetrix Human Clariom D Assay PSR1900167557.hg.1 TC1900008649.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900167557.hg.1 ENST00000396477.4 Ensembl
Affymetrix Human Clariom D Assay PSR1900167557.hg.1 NM_001278392.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0