This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs00999721_m1
See other MSS51 GE Assays ›
Gene Symbol:
MSS51
Gene Name:
MSS51 mitochondrial translational activator
Gene Aliases:
ZMYND17
Chromosome Location:
Chr.10: 73423579 - 73433561 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs00999721_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: MSS51
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA AK309870.1 - 2 - 3 105 97

Back To Top

More Information


Set Membership:

Amplicon length between 86 and 100 Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000299432.6 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000372912.1 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 ENST00000487126.5 Ensembl
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq
Affymetrix Human Clariom D Assay TC1000011023.hg.1 NM_001024593.1 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-4w7vk:80/100.66.78.22:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0