This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01001184_g1
See other SLC7A5 GE Assays ›
Gene Symbol:
SLC7A5
Gene Name:
solute carrier family 7 member 5
Gene Aliases:
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1
Chromosome Location:
Chr.16: 87830022 - 87869499 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01001184_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003486.5 NP_003477.4 3 - 4 843 72
XM_006721286.3 XP_006721349.2 3 - 4 486 72
XM_017023735.1 XP_016879224.1 3 - 4 848 72
XM_017023736.1 XP_016879225.1 2 - 3 722 72
XM_017023737.1 XP_016879226.1 3 - 4 435 72
GenBank mRNA AB017908.1 - 3 - 4 811 72
AB018009.1 - 3 - 4 836 72
AB018542.1 - 3 - 4 786 72
AF077866.1 - 3 - 4 823 72
AF104032.1 - 3 - 4 837 72
BC039692.2 - 3 - 4 868 5551612 72
BC042600.1 - 3 - 4 837 4470593 72
M80244.1 - 3 - 4 283 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011186.hg
This assay targets the same location as HTA transcript cluster(s) TC16001335.hg

Additional Information:

This assay is in the 5'UTR region of the M80244.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A5

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
transport
amino acid transport
nervous system development
neutral amino acid transport
L-amino acid transport
cell differentiation
leukocyte migration
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity
peptide antigen binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vnb2r:80/100.66.79.197:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0