This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01001188_m1
See other SLC7A5 GE Assays ›
Gene Symbol:
SLC7A5
Gene Name:
solute carrier family 7 member 5
Gene Aliases:
4F2LC, CD98, D16S469E, E16, LAT1, MPE16, hLAT1
Chromosome Location:
Chr.16: 87830022 - 87869499 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01001188_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A5
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_003486.5 NP_003477.4 7 - 8 1212 108
XM_006721286.3 XP_006721349.2 7 - 8 855 108
XM_017023735.1 XP_016879224.1 7 - 8 1217 108
XM_017023736.1 XP_016879225.1 6 - 7 1091 108
XM_017023737.1 XP_016879226.1 7 - 8 804 108
GenBank mRNA AB017908.1 - 7 - 8 1180 108
AB018009.1 - 7 - 8 1205 108
AB018542.1 - 7 - 8 1155 108
AF077866.1 - 7 - 8 1192 108
AF104032.1 - 7 - 8 1206 108
BC039692.2 - 7 - 8 1237 5551612 108
BC042600.1 - 7 - 8 1206 4470593 108
M80244.1 - 7 - 8 652 108

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011186.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16001335.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A5

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

cellular amino acid metabolic process
transport
amino acid transport
nervous system development
neutral amino acid transport
L-amino acid transport
cell differentiation
leukocyte migration
L-alpha-amino acid transmembrane transport
amino acid transmembrane transporter activity
neutral amino acid transmembrane transporter activity
L-amino acid transmembrane transporter activity
antiporter activity
peptide antigen binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000261622.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000563489.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 ENST00000565644.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600011186.hg.1 NM_003486.6 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x88k6:80/100.66.75.98:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0