This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01003686_m1
See other UBE2J2 GE Assays ›
Gene Symbol:
UBE2J2
Gene Name:
ubiquitin conjugating enzyme E2 J2
Gene Aliases:
NCUBE-2, NCUBE2, PRO2121
Chromosome Location:
Chr.1: 1253912 - 1273854 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01003686_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UBE2J2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011540616.2 XP_011538918.1 4 - 5 601 113
XM_011540617.2 XP_011538919.1 3 - 4 368 113
XM_017000238.1 XP_016855727.1 4 - 5 472 113
GenBank mRNA AL555293.3 - 4 - 5 485 113

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the XM_011540616.2, XM_011540617.2, XM_017000238.1 transcripts.

Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

protein polyubiquitination
response to unfolded protein
ER-associated ubiquitin-dependent protein catabolic process
positive regulation of protein targeting to mitochondrion
ATP binding
ubiquitin protein ligase binding
ubiquitin protein ligase activity
ubiquitin conjugating enzyme activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-d7bxm:80/100.66.76.204:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0