This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01006342_m1
See other TRAK1 GE Assays ›
Gene Symbol:
TRAK1
Gene Name:
trafficking kinesin protein 1
Gene Aliases:
MILT1, OIP106
Chromosome Location:
Chr.3: 42013802 - 42225890 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01006342_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TRAK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001042646.2 NP_001036111.1 14 - 15 2183 72
XM_005264958.2 XP_005265015.1 13 - 14 2046 72
XM_005264960.3 XP_005265017.1 14 - 15 1991 72
GenBank mRNA AB028965.1 - 14 - 15 2183 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013812.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03000217.hg.1

Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

membrane traffic protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of transcription from RNA polymerase II promoter
protein O-linked glycosylation
protein targeting
endosome to lysosome transport
protein binding
GABA receptor binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 TC03000217.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 ENST00000327628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 ENST00000341421.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 ENST00000396175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 ENST00000487159.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 NM_001042646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004352.hg.1 NM_014965.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004358.hg.1 TC03000217.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03004358.hg.1 ENST00000327628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004358.hg.1 ENST00000396175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004358.hg.1 ENST00000487159.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004358.hg.1 ENST00000543338.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004358.hg.1 NM_001042646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 TC03000217.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 ENST00000327628.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 ENST00000341421.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 ENST00000396175.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 ENST00000487159.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 ENST00000543338.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 NM_001042646.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR03004351.hg.1 NM_014965.4 Refseq
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 ENST00000327628.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 ENST00000396175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 NM_001042646.2 Refseq
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 ENST00000327628.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 ENST00000396175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 NM_001042646.2 Refseq
Affymetrix Human Clariom D Assay PSR0300203794.hg.1 TC0300013812.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000327628.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000341421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000396175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000613405.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_001042646.2 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_001265608.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_001265609.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_014965.4 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000327628.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000341421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000396175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 ENST00000613405.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_001042646.2 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_001265608.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_001265609.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 NM_014965.4 Refseq
Affymetrix Human Clariom D Assay PSR0300203787.hg.1 TC0300013812.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000327628.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000341421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000396175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000613405.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_001042646.2 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_001265608.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_001265609.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_014965.4 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000327628.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000341421.7 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000396175.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 ENST00000613405.4 Ensembl
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_001042646.2 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_001265608.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_001265609.1 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 NM_014965.4 Refseq
Affymetrix Human Clariom D Assay PSR0300203788.hg.1 TC0300013812.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-x8dcn:80/100.66.79.134:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0