This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01006873_g1
See other TNK2 GE Assays ›
Gene Symbol:
TNK2
Gene Name:
tyrosine kinase non receptor 2
Gene Aliases:
ACK, ACK-1, ACK1, p21cdc42Hs
Chromosome Location:
Chr.3: 195863364 - 195909009 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01006873_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TNK2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005781.4 NP_005772.3 12 - 13 3434 97
XM_011512318.2 XP_011510620.2 13 - 14 3209 97
XM_017005509.1 XP_016860998.1 12 - 13 3164 97
GenBank mRNA L13738.2 - 12 - 13 3503 97

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300013670.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03002152.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Protein Kinase Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TNK2

Gene Ontology Categories:

Function(s) Process(es)

endocytosis
cell surface receptor signaling pathway
transmembrane receptor protein tyrosine kinase signaling pathway
small GTPase mediated signal transduction
phosphorylation
cell migration
cell differentiation
negative regulation of GTPase activity
peptidyl-tyrosine autophosphorylation
regulation of cell proliferation
innate immune response
positive regulation of peptidyl-tyrosine phosphorylation
regulation of clathrin-mediated endocytosis
protein serine/threonine kinase activity
protein serine/threonine/tyrosine kinase activity
protein tyrosine kinase activity
non-membrane spanning protein tyrosine kinase activity
GTPase inhibitor activity
epidermal growth factor receptor binding
protein binding
ATP binding
identical protein binding
metal ion binding
WW domain binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03037969.hg.1 TC03002152.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03037969.hg.1 ENST00000333602.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03037969.hg.1 ENST00000392400.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03037969.hg.1 NM_005781.4 Refseq
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 ENST00000333602.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 ENST00000416152.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 ENST00000420716.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 NM_005781.4 Refseq
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 ENST00000333602.10 Ensembl
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 ENST00000416152.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 ENST00000420716.2 Ensembl
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 NM_005781.4 Refseq
Affymetrix Human Clariom D Assay PSR0300202019.hg.1 TC0300013670.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-wf7gw:80/100.66.73.223:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0