| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
ENST00000264917.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
ENST00000333194.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
ENST00000340978.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
ENST00000342343.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
ENST00000346042.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
ENST00000505283.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
NM_001029851.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
NM_001029852.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
NM_001029853.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
NM_001029854.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
NM_003719.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05005361.hg.1 |
TC05000381.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000264917.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000333194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000340978.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000342343.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000346042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000505283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029853.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029854.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_003719.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000264917.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000333194.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000340978.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000342343.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000346042.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
ENST00000505283.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029851.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029852.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029853.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_001029854.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
NM_003719.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500153779.hg.1 |
TC0500007848.hg.1 |
Clariom D TC |