This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01008432_m1
See other EEF1G GE Assays ›
Gene Symbol:
EEF1G
Gene Name:
eukaryotic translation elongation factor 1 gamma
Gene Aliases:
EF1G, GIG35
Chromosome Location:
Chr.11: 62559601 - 62573988 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01008432_m1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF1G
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001404.4 NP_001395.1 1 - 2 158 84
GenBank mRNA BQ947806.1 - 1 - 2 158 84

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100013170.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003483.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001404.4 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF1G

Gene Ontology Categories:

Function(s) Process(es)

translational elongation
glutathione metabolic process
response to virus
cell-cell adhesion
translation elongation factor activity
glutathione transferase activity
protein binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11021651.hg.1 TC11003483.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11021651.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021651.hg.1 ENST00000378019.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021651.hg.1 ENST00000424909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021651.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021651.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11021647.hg.1 TC11003483.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11021647.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021647.hg.1 ENST00000378019.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021647.hg.1 ENST00000424909.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021647.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021647.hg.1 NM_001404.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11021649.hg.1 TC11003483.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR11021649.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021649.hg.1 ENST00000378019.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021649.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11021649.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204217.hg.1 TC1100013170.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204220.hg.1 TC1100013170.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204222.hg.1 TC1100013170.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204223.hg.1 TC1100013170.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000329251.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000524420.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000525340.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 ENST00000532986.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 NM_001404.4 Refseq
Affymetrix Human Clariom D Assay PSR1100204218.hg.1 TC1100013170.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-w8bxd:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0