This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01009052_m1
See other TCF7L2 GE Assays ›
Gene Symbol:
TCF7L2
Gene Name:
transcription factor 7 like 2
Gene Aliases:
TCF-4, TCF4
Chromosome Location:
Chr.10: 112950219 - 113167678 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01009052_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TCF7L2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001146285.1 NP_001139757.1 12 - 13 1829 75
NM_030756.4 NP_110383.2 13 - 14 1880 75
XM_005270071.1 XP_005270128.1 15 - 16 2139 75
XM_005270075.1 XP_005270132.1 15 - 16 2124 75
XM_005270077.1 XP_005270134.1 14 - 15 2088 75
XM_005270079.1 XP_005270136.1 14 - 15 2073 75
XM_005270080.1 XP_005270137.1 14 - 15 2070 75
XM_005270084.1 XP_005270141.1 14 - 15 1980 75
XM_005270086.1 XP_005270143.1 13 - 14 1926 75
XM_005270088.1 XP_005270145.1 13 - 14 1899 75
XM_017016593.1 XP_016872082.1 13 - 14 1929 75
GenBank mRNA AB451266.1 - 13 - 14 1373 75
AK225809.1 - 12 - 13 1833 75
CR536574.1 - 13 - 14 1373 75
FJ010173.1 - 12 - 13 1268 75
HM352845.1 - 14 - 15 1502 75
HM352848.1 - 13 - 14 1373 75
HM352850.1 - 14 - 15 1442 75
Y11306.2 - 13 - 14 1680 75

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 3' Most Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TCF7L2

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
blood vessel development
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
cell cycle arrest
Wnt signaling pathway, calcium modulating pathway
cell proliferation
response to glucose
positive regulation of heparan sulfate proteoglycan biosynthetic process
pancreas development
positive regulation of insulin secretion
positive regulation of protein binding
regulation of hormone metabolic process
glucose homeostasis
negative regulation of sequence-specific DNA binding transcription factor activity
maintenance of DNA repeat elements
canonical Wnt signaling pathway involved in positive regulation of epithelial to mesenchymal transition
fat cell differentiation
negative regulation of transcription, DNA-templated
positive regulation of transcription from RNA polymerase II promoter
positive regulation of protein export from nucleus
myoblast fate commitment
regulation of smooth muscle cell proliferation
positive regulation of protein kinase B signaling
canonical Wnt signaling pathway
negative regulation of canonical Wnt signaling pathway
beta-catenin-TCF complex assembly
negative regulation of type B pancreatic cell apoptotic process
negative regulation of extrinsic apoptotic signaling pathway
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II repressing transcription factor binding
transcription factor activity, sequence-specific DNA binding
protein binding
beta-catenin binding
transcription factor binding
protein kinase binding
nuclear hormone receptor binding
sequence-specific DNA binding
transcription regulatory region DNA binding
gamma-catenin binding
armadillo repeat domain binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-5thf6:80/100.66.79.225:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0