This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01009177_m1
See other DCHS2 GE Assays ›
Gene Symbol:
DCHS2
Gene Name:
dachsous cadherin-related 2
Gene Aliases:
CDH27, CDHJ, CDHR7, PCDH23, PCDHJ
Chromosome Location:
Chr.4: 154234375 - 154491778 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01009177_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DCHS2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017639.3 NP_060109.2 22 - 23 5710 128
XM_011532045.1 XP_011530347.1 17 - 18 7498 128
XM_011532046.2 XP_011530348.1 16 - 17 5154 128
XM_011532049.2 XP_011530351.1 9 - 10 3004 128
GenBank mRNA AK095103.1 - 5 - 6 1047 128
AY354497.1 - 22 - 23 5710 128
BX647835.1 - 5 - 6 686 128

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0400012178.hg.1
This assay targets the same location as HTA transcript cluster(s) TC04001659.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DCHS2

Panther Classification:

Molecular Function -

cadherin

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
biological_process
molecular_function
calcium ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04023056.hg.1 ENST00000357232.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04023056.hg.1 NM_017639.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04023056.hg.1 TC04001659.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 ENST00000357232.8 Ensembl
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 ENST00000623607.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 NM_017639.3 Refseq
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 ENST00000357232.8 Ensembl
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 ENST00000623607.3 Ensembl
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 NM_017639.3 Refseq
Affymetrix Human Clariom D Assay PSR0400183566.hg.1 TC0400012178.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline