| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028869.hg.1 |
ENST00000326621.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028869.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028869.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028869.hg.1 |
TC11003495.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028868.hg.1 |
ENST00000326621.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028868.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028868.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028868.hg.1 |
TC11003495.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028870.hg.1 |
ENST00000326621.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028870.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028870.hg.1 |
ENST00000442070.2 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028870.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028870.hg.1 |
TC11003495.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR11028870.hg.1 |
TC11003496.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100197487.hg.1 |
TC1100012685.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100197488.hg.1 |
TC1100012685.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1100197489.hg.1 |
TC1100012685.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000326621.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
ENST00000403470.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1100012685.hg.1 |
NM_014312.4 |
Refseq |