This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01009783_g1
See other VSIG2 GE Assays ›
Gene Symbol:
VSIG2
Gene Name:
V-set and immunoglobulin domain containing 2
Gene Aliases:
2210413P10Rik, CTH, CTXL
Chromosome Location:
Chr.11: 124747474 - 124752213 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01009783_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: VSIG2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_014312.3 NP_055127.2 2 - 3 290 65
GenBank mRNA AK000460.1 - 2 - 3 271 65
AY358897.1 - 2 - 3 315 65
BC007313.2 - 2 - 3 290 2987937 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012685.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11003495.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene VSIG2

Panther Classification:

Molecular Function -

defense/immunity protein immunoglobulin receptor superfamily

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11028867.hg.1 ENST00000326621.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11028867.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11028867.hg.1 NM_014312.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11028867.hg.1 TC11003495.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay PSR1100197486.hg.1 TC1100012685.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000326621.9 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 ENST00000403470.1 Ensembl
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq
Affymetrix Human Clariom D Assay TC1100012685.hg.1 NM_014312.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-744bc48644-454p7:80/100.66.72.101:80.
git-commit: 747bde55a712f6f97bf7760408d445eefba4e16f
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.0-Offline