This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01009788_m1
See other OPCML GE Assays ›
Gene Symbol:
OPCML
Gene Name:
opioid binding protein/cell adhesion molecule like
Gene Aliases:
IGLON1, OBCAM, OPCM
Chromosome Location:
Chr.11: 132403361 - 133532983 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01009788_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: OPCML
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001319106.1 NP_001306035.1 3 - 4 1074 69
XM_006718846.2 XP_006718909.1 4 - 5 1022 69
GenBank mRNA AK314077.1 - 3 - 4 1074 69
BC074742.2 - 3 - 4 556 30915458 69
BC143945.1 - 3 - 4 552 9052463 69

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100012882.hg.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

immunoglobulin

Gene Ontology Categories:

Function(s)

cell adhesion
neuron recognition

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000331898.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000374778.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000524381.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000529038.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000541867.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000612177.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 NM_001012393.2 Refseq
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 NM_002545.4 Refseq
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000331898.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000374778.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000524381.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000529038.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000541867.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 ENST00000612177.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 NM_001012393.2 Refseq
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 NM_002545.4 Refseq
Affymetrix Human Clariom D Assay PSR1100198770.hg.1 TC1100012882.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 ENST00000331898.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 ENST00000541867.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 ENST00000612177.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 NM_002545.4 Refseq
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 ENST00000331898.11 Ensembl
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 ENST00000541867.5 Ensembl
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 ENST00000612177.4 Ensembl
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 NM_002545.4 Refseq
Affymetrix Human Clariom D Assay PSR1100198777.hg.1 TC1100012882.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline