This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01010141_m1
See other SDK1 GE Assays ›
Gene Symbol:
SDK1
Gene Name:
sidekick cell adhesion molecule 1
Gene Aliases:
-
Chromosome Location:
Chr.7: 3301448 - 4269000 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01010141_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SDK1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001079653.1 NP_001073121.1 2 - 3 509 72
NM_152744.3 NP_689957.3 28 - 29 4364 72
XM_011515188.1 XP_011513490.1 22 - 23 3202 72
XM_011515189.1 XP_011513491.1 22 - 23 3202 72
XM_011515190.1 XP_011513492.1 22 - 23 3202 72
XM_017011837.1 XP_016867326.1 22 - 23 3202 72
XM_017011838.1 XP_016867327.1 22 - 23 3202 72
XM_017011839.1 XP_016867328.1 22 - 23 3202 72
XM_017011840.1 XP_016867329.1 22 - 23 3202 72
GenBank mRNA AK074083.1 - 14 - 15 2060 72
AL833354.1 - 2 - 3 506 72
AY310398.1 - 28 - 29 4364 72
DB065853.1 - 2 - 3 505 72

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700006583.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000040.hg.1

Additional Information:

This assay is in the 5'UTR region of the NM_001079653.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SDK1

Gene Ontology Categories:

Function(s) Process(es)

homophilic cell adhesion via plasma membrane adhesion molecules
synapse assembly
retina layer formation
behavioral response to cocaine
regulation of dendritic spine development
identical protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07000621.hg.1 TC07000040.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07000621.hg.1 ENST00000389531.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000621.hg.1 ENST00000404826.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000621.hg.1 ENST00000446104.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000621.hg.1 NM_001079653.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07000621.hg.1 NM_152744.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000389531.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000404826.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000476701.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000615806.4 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 NM_001079653.1 Refseq
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 NM_152744.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000389531.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000404826.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000476701.5 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 ENST00000615806.4 Ensembl
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 NM_001079653.1 Refseq
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 NM_152744.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145247.hg.1 TC0700006583.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0