This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01010435_g1
See other LINC00400 GE Assays ›
Gene Symbol:
LINC00400
Gene Name:
long intergenic non-protein coding RNA 400
Gene Aliases:
-
Chromosome Location:
Chr.13: 43113165 - 43159466 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01010435_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: LINC00400
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_047013.1 2 - 3 489 128
GenBank mRNA AV711458.1 - 2 - 3 489 128

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Detect Genomic DNA Homo sapiens Probe spans exons

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1300009955.hg.1 ENST00000425609.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1300009955.hg.1 ENST00000425609.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009955.hg.1 ENST00000425609.1 Ensembl
Affymetrix Human Clariom D Assay TC1300009955.hg.1 ENST00000425609.1 Ensembl

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-xql92:80/100.66.76.113:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline