This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01011312_g1
See other RBM11 GE Assays ›
Gene Symbol:
RBM11
Gene Name:
RNA binding motif protein 11
Gene Aliases:
-
Chromosome Location:
Chr.21: 14216107 - 14228372 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01011312_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM11
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001320602.1 NP_001307531.1 3 - 4 384 93
NM_144770.4 NP_658983.3 3 - 4 384 93
NR_135308.1 2 - 3 221 93
NR_135309.1 2 - 3 221 93
NR_135310.1 3 - 4 394 93
NR_135311.1 3 - 4 394 93
XM_005260997.4 XP_005261054.1 3 - 4 407 93
XM_017028386.1 XP_016883875.1 3 - 4 539 93
XM_017028387.1 XP_016883876.1 3 - 4 546 93
GenBank mRNA AF519623.1 - 2 - 3 185 93
AK091331.1 - 3 - 4 366 93
AY077695.1 - 3 - 4 315 93
BC030196.1 - 3 - 4 336 5199053 93
CB153254.1 - 3 - 4 394 93
CN430785.1 - 3 - 4 380 93

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC2100006585.hg
This assay targets the same location as HTA transcript cluster(s) TC21000025.hg

Additional Information:

This assay is in the 5'UTR region of the AK091331.1,AY077695.1,BC030196.1,XM_017028386.1,XM_017028387.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM11

Panther Classification:

Molecular Function -

nucleic acid metabolism protein

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
multicellular organism development
RNA splicing
cell differentiation
cellular response to oxidative stress
nucleotide binding
protein binding
poly(U) RNA binding
protein homodimerization activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000400577.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000461088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000461088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000461088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000461088.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468643.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000468788.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000475864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000475864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000475864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000475864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000475864.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 ENST00000495055.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC2100006585.hg.1 NM_144770.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0