This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01012127_g1
See other SLC7A10 GE Assays ›
Gene Symbol:
SLC7A10
Gene Name:
solute carrier family 7 member 10
Gene Aliases:
ASC1, HASC-1, asc-1
Chromosome Location:
Chr.19: 33208664 - 33225850 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01012127_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_019849.2 NP_062823.1 7 - 8 1164 101
GenBank mRNA AB037670.1 - 7 - 8 1116 101
AJ277731.1 - 7 - 8 1125 101
AK316594.1 - 7 - 8 1036 101
BC035627.1 - 7 - 8 1107 5200601 101

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900010379.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001395.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Transporter Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC7A10

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
leukocyte migration
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR19019237.hg.1 ENST00000253188.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19019237.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19019237.hg.1 TC19001395.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR19019236.hg.1 ENST00000253188.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19019236.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19019236.hg.1 TC19001395.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 TC1900010379.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900185066.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 TC1900010379.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900185067.hg.1 NM_019849.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline