This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01012841_m1
See other EEF2 GE Assays ›
Gene Symbol:
EEF2
Gene Name:
eukaryotic translation elongation factor 2
Gene Aliases:
EEF-2, EF-2, EF2, SCA26
Chromosome Location:
Chr.19: 3976056 - 3985463 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01012841_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EEF2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001961.3 NP_001952.1 2 - 3 304 88
GenBank mRNA AK297398.1 - 2 - 3 304 88
AK298498.1 - 2 - 3 304 88
AK299225.1 - 2 - 3 300 88
AU118535.1 - 2 - 3 304 88
AY942181.1 - 1 - 2 221 88
BC006547.2 - 2 - 3 289 88
BC126259.1 - 2 - 3 273 8991975 88
BC136313.1 - 2 - 3 273 9020300 88
X51466.1 - 1 - 2 221 88
Z11692.1 - 1 - 2 221 88

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1900009316.hg.1
This assay targets the same location as HTA transcript cluster(s) TC19001054.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EEF2

Panther Classification:

Molecular Function -

translation elongation factor

Gene Ontology Categories:

Function(s) Process(es)

hematopoietic progenitor cell differentiation
response to ischemia
skeletal muscle contraction
translational elongation
aging
glial cell proliferation
response to fluoxetine
peptidyl-diphthamide biosynthetic process from peptidyl-histidine
response to estradiol
response to endoplasmic reticulum stress
skeletal muscle cell differentiation
response to hydrogen peroxide
response to ethanol
positive regulation of translation
response to folic acid
cell-cell adhesion
cellular response to brain-derived neurotrophic factor stimulus
positive regulation of cytoplasmic translation
p53 binding
translation elongation factor activity
GTPase activity
protein binding
GTP binding
5S rRNA binding
protein kinase binding
ribosome binding
poly(A) RNA binding
actin filament binding
cadherin binding involved in cell-cell adhesion

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000384048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000384048.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000596417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000596417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000596417.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NR_002602.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900009316.hg.1 NR_002602.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19014418.hg.1 ENST00000309311.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19014418.hg.1 ENST00000543343.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR19014418.hg.1 NM_001961.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR19014418.hg.1 TC19001054.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000384048.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000384048.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000594885.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000596417.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000596417.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000596417.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598182.5 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000598436.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 ENST00000600794.1 Ensembl
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NR_002602.1 Refseq
Affymetrix Human Clariom D Assay TC1900009316.hg.1 NR_002602.1 Refseq
Affymetrix Human Clariom D Assay PSR1900174401.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900174401.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174401.hg.1 TC1900009316.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900174401.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900174401.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 NM_001961.3 Refseq
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 TC1900009316.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 ENST00000309311.6 Ensembl
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 ENST00000600720.1 Ensembl
Affymetrix Human Clariom D Assay PSR1900174402.hg.1 NM_001961.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0