This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01013116_g1
See other STAT2 GE Assays ›
Gene Symbol:
STAT2
Gene Name:
signal transducer and activator of transcription 2
Gene Aliases:
IMD44, ISGF-3, P113, STAT113
Chromosome Location:
Chr.12: 56341597 - 56360253 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01013116_g1
Size
Availability Inventoried
Catalog # 4331182
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: STAT2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_005419.3 NP_005410.1 11 - 12 1300 76
NM_198332.1 NP_938146.1 11 - 12 1288 76
XM_005269111.3 XP_005269168.2 11 - 12 1208 76
XM_011538697.2 XP_011536999.1 11 - 12 1208 76
XM_011538698.2 XP_011537000.1 11 - 12 1169 76
XM_011538699.2 XP_011537001.1 11 - 12 1208 76
XM_011538700.2 XP_011537002.1 5 - 6 606 76
XM_017019904.1 XP_016875393.1 10 - 11 1667 76
GenBank mRNA AK296939.1 - 11 - 12 1161 76
AK296940.1 - 11 - 12 1161 76
BC006092.1 - 10 - 11 1087 76
BC051284.1 - 10 - 11 1120 6460486 76
M97934.1 - 11 - 12 1134 76

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Inventoried Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene STAT2

Panther Classification:

Molecular Function -

DNA-binding transcription factor

Gene Ontology Categories:

Function(s) Process(es)

regulation of protein phosphorylation
positive regulation of defense response to virus by host
transcription, DNA-templated
regulation of transcription from RNA polymerase II promoter
JAK-STAT cascade
defense response to virus
type I interferon signaling pathway
regulation of type I interferon-mediated signaling pathway
regulation of mitochondrial fission
mitophagy in response to mitochondrial depolarization
xenophagy
DNA binding
transcription factor activity, sequence-specific DNA binding
signal transducer activity
protein binding
identical protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-97tfg:80/100.66.76.242:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline