This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01013869_m1
See other NSUN6 GE Assays ›
Gene Symbol:
NSUN6
Gene Name:
NOP2/Sun RNA methyltransferase family member 6
Gene Aliases:
4933414E04Rik, ARL5B-AS1, NOPD1
Chromosome Location:
Chr.10: 18512582 - 18659264 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01013869_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NSUN6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182543.3 NP_872349.1 6 - 7 1094 104
XM_011519383.1 XP_011517685.1 6 - 7 1093 104
XM_011519384.1 XP_011517686.1 8 - 9 1078 104
XM_011519385.1 XP_011517687.1 7 - 8 767 104
XM_011519386.2 XP_011517688.1 7 - 8 820 104
XM_011519387.2 XP_011517689.1 5 - 6 4331 104
XM_011519389.2 XP_011517691.1 4 - 5 445 104
XM_017015906.1 XP_016871395.1 6 - 7 1081 104
XM_017015907.1 XP_016871396.1 7 - 8 1003 104
XM_017015908.1 XP_016871397.1 7 - 8 808 104
XM_017015909.1 XP_016871398.1 5 - 6 4319 104
XM_017015910.1 XP_016871399.1 5 - 6 4407 104
XM_017015911.1 XP_016871400.1 5 - 6 4394 104
XM_017015912.1 XP_016871401.1 4 - 5 531 104
XM_017015913.1 XP_016871402.1 7 - 8 4546 104
XM_017015914.1 XP_016871403.1 4 - 5 4238 104
GenBank mRNA AK074323.1 - 6 - 7 1047 104
BC035778.1 - 6 - 7 1080 5590372 104

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012528.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10002961.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NSUN6

Gene Ontology Categories:

Function(s) Process(es)

RNA methylation
tRNA methylation
tRNA binding
RNA methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR10013657.hg.1 TC10002961.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR10013657.hg.1 ENST00000377304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013657.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186733.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000186733.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186733.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000186733.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186733.hg.1 TC1000012528.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0