This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01013871_m1
See other NSUN6 GE Assays ›
Gene Symbol:
NSUN6
Gene Name:
NOP2/Sun RNA methyltransferase family member 6
Gene Aliases:
4933414E04Rik, ARL5B-AS1, NOPD1
Chromosome Location:
Chr.10: 18512582 - 18659264 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01013871_m1
Best Coverage
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: NSUN6
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_182543.3 NP_872349.1 9 - 10 1508 62
XM_011519383.1 XP_011517685.1 9 - 10 1507 62
XM_011519384.1 XP_011517686.1 11 - 12 1492 62
XM_011519385.1 XP_011517687.1 10 - 11 1181 62
XM_011519386.2 XP_011517688.1 10 - 11 1234 62
XM_011519387.2 XP_011517689.1 8 - 9 4745 62
XM_011519389.2 XP_011517691.1 7 - 8 859 62
XM_017015906.1 XP_016871395.1 9 - 10 1495 62
XM_017015907.1 XP_016871396.1 10 - 11 1417 62
XM_017015908.1 XP_016871397.1 10 - 11 1222 62
XM_017015909.1 XP_016871398.1 8 - 9 4733 62
XM_017015910.1 XP_016871399.1 8 - 9 4821 62
XM_017015911.1 XP_016871400.1 8 - 9 4808 62
XM_017015912.1 XP_016871401.1 7 - 8 945 62
XM_017015913.1 XP_016871402.1 10 - 11 4960 62
XM_017015914.1 XP_016871403.1 7 - 8 4652 62
GenBank mRNA AK074323.1 - 9 - 10 1461 62
BC035778.1 - 9 - 10 1494 5590372 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1000012528.hg.1
This assay targets the same location as HTA transcript cluster(s) TC10002961.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Recommended Assay Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene NSUN6

Gene Ontology Categories:

Function(s) Process(es)

RNA methylation
tRNA methylation
tRNA binding
RNA methyltransferase activity
S-adenosylmethionine-dependent methyltransferase activity
tRNA (cytosine-5-)-methyltransferase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10013653.hg.1 ENST00000377304.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013653.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR10013653.hg.1 NM_182543.4 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR10013653.hg.1 TC10002961.hg.1 HTA TC
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186729.hg.1 TC1000012528.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1000186731.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000186731.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186731.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay PSR1000186731.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay PSR1000186731.hg.1 TC1000012528.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000377304.6 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000425669.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000433526.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000436485.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 ENST00000493816.1 Ensembl
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq
Affymetrix Human Clariom D Assay TC1000012528.hg.1 NM_182543.4 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-6b469b8bb7-msbq7:80/100.66.78.64:80.
git-commit: a334af76dff23450325448aedefe62379591458a
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.48.1-2026.04.16-1.0