This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01014627_g1
See other UNK GE Assays ›
Gene Symbol:
UNK
Gene Name:
unkempt family zinc finger
Gene Aliases:
UNKEMPT, ZC3H5, ZC3HDC5
Chromosome Location:
Chr.17: 75784771 - 75825805 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01014627_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UNK
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001080419.2 NP_001073888.2 10 - 11 1417 116
XM_006722145.3 XP_006722208.1 13 - 14 5762 116
XM_006722148.2 XP_006722211.1 11 - 12 1558 116
XM_011525400.1 XP_011523702.1 11 - 12 1486 116
XM_011525401.2 XP_011523703.1 5 - 6 574 116
XM_017025247.1 XP_016880736.1 13 - 14 5798 116
XM_017025248.1 XP_016880737.1 12 - 13 5700 116
XM_017025249.1 XP_016880738.1 12 - 13 1843 116
XM_017025250.1 XP_016880739.1 11 - 12 1745 116
GenBank mRNA AB051540.1 - 10 - 11 1400 116
BC053362.1 - 10 - 11 1400 116

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1700008923.hg.1
This assay targets the same location as HTA transcript cluster(s) TC17000862.hg.1

Set Membership:

Amplicon length greater than or equal to 101 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

neuron migration
cell morphogenesis involved in neuron differentiation
negative regulation of cytoplasmic translation
poly(A) RNA binding
metal ion binding
mRNA CDS binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587501.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587501.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000587501.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000589790.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592074.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17011244.hg.1 ENST00000293218.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR17011244.hg.1 NM_001080419.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR17011244.hg.1 TC17000862.hg.1 HTA TC
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586217.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000586527.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587258.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587501.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587501.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000587501.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589790.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589790.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000589790.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592074.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592074.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592074.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 ENST00000592629.1 Ensembl
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Clariom D Assay TC1700008923.hg.1 NR_038131.1 Refseq
Affymetrix Human Clariom D Assay PSR1700167197.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700167197.hg.1 NM_001080419.2 Refseq
Affymetrix Human Clariom D Assay PSR1700167197.hg.1 TC1700008923.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR1700167197.hg.1 ENST00000589666.5 Ensembl
Affymetrix Human Clariom D Assay PSR1700167197.hg.1 NM_001080419.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-zvjvc:80/100.66.72.70:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline