Assay Details
| Gene Symbol: UNK | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001080419.2 | NP_001073888.2 | 7 - 8 | 1006 | 55 | |
| XM_006722145.3 | XP_006722208.1 | 10 - 11 | 5387 | 55 | ||
| XM_006722148.2 | XP_006722211.1 | 8 - 9 | 1147 | 55 | ||
| XM_011525400.1 | XP_011523702.1 | 8 - 9 | 1075 | 55 | ||
| XM_011525401.2 | XP_011523703.1 | 2 - 3 | 163 | 55 | ||
| XM_017025247.1 | XP_016880736.1 | 10 - 11 | 5387 | 55 | ||
| XM_017025248.1 | XP_016880737.1 | 9 - 10 | 5289 | 55 | ||
| XM_017025249.1 | XP_016880738.1 | 9 - 10 | 1432 | 55 | ||
| XM_017025250.1 | XP_016880739.1 | 8 - 9 | 1334 | 55 | ||
| GenBank mRNA | AB051540.1 | - | 7 - 8 | 989 | 55 | |
| BC053362.1 | - | 7 - 8 | 989 | 55 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17000862.hg.1
Set Membership: |
|
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR17011241.hg.1 | ENST00000293218.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011241.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011241.hg.1 | TC17000862.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011240.hg.1 | ENST00000293218.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011240.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17011240.hg.1 | TC17000862.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700167194.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167194.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167194.hg.1 | TC1700008923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700167194.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167194.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000586527.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587258.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000587501.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000589790.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592074.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | ENST00000592629.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700008923.hg.1 | NR_038131.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167193.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167193.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167193.hg.1 | TC1700008923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700167193.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167193.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | NM_001080419.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | TC1700008923.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | ENST00000586217.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | ENST00000589666.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700167192.hg.1 | NM_001080419.2 | Refseq |

UNK
Genomic Map