This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01015334_m1
See other EIF3M GE Assays ›
Gene Symbol:
EIF3M
Gene Name:
eukaryotic translation initiation factor 3 subunit M
Gene Aliases:
B5, GA17, PCID1, TANGO7, hfl-B5
Chromosome Location:
Chr.11: 32583767 - 32602873 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01015334_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: EIF3M
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001307929.1 NP_001294858.1 6 - 7 667 83
NM_006360.5 NP_006351.2 9 - 10 1063 83
XM_011519840.2 XP_011518142.1 9 - 10 1087 83
GenBank mRNA AF064603.1 - 9 - 10 993 83
AF277183.1 - 5 - 6 432 83
AK131064.1 - 9 - 10 1017 83
AK222564.1 - 9 - 10 982 83
AK292139.1 - 9 - 10 1005 83
AK304378.1 - 6 - 7 603 83
AK312512.1 - 9 - 10 1032 83
AV713889.1 - 6 - 7 584 83
AY769947.1 - 9 - 10 942 83
BC019103.2 - 9 - 10 985 5021401 83
BC020458.1 - 5 - 6 454 83
BC051292.1 - 9 - 10 987 3607828 83
CR450300.1 - 9 - 10 942 83

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene EIF3M

Gene Ontology Categories:

Function(s) Process(es)

formation of translation preinitiation complex
cytoplasmic translational initiation
translational initiation
regulation of translational initiation
translation initiation factor activity
protein binding
translation initiation factor binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-jmd5k:80/100.66.78.41:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0