This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01015942_m1
See other BCO1 GE Assays ›
Gene Symbol:
BCO1
Gene Name:
beta-carotene oxygenase 1
Gene Aliases:
BCDO, BCDO1, BCMO, BCMO1, BCO
Chromosome Location:
Chr.16: 81238642 - 81291142 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01015942_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017429.2 NP_059125.2 3 - 4 535 63
XM_011523109.1 XP_011521411.1 3 - 4 584 63
XM_017023286.1 XP_016878775.1 3 - 4 584 63
XM_017023287.1 XP_016878776.1 3 - 4 584 63
XM_017023288.1 XP_016878777.1 3 - 4 584 63
GenBank mRNA AF294900.1 - 3 - 4 535 63
AK001592.1 - 3 - 4 535 63
BC126210.1 - 3 - 4 437 8991926 63
BC126212.1 - 3 - 4 437 8991928 63

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008577.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000637.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BCO1

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
vitamin A biosynthetic process
retinol metabolic process
retinal metabolic process
oxidation-reduction process
beta-carotene metabolic process
beta-carotene 15,15'-monooxygenase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16009011.hg.1 ENST00000258168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009011.hg.1 ENST00000425577.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009011.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16009011.hg.1 TC16000637.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay PSR1600163998.hg.1 TC1600008577.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-tks2p:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0