This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01015946_m1
See other BCO1 GE Assays ›
Gene Symbol:
BCO1
Gene Name:
beta-carotene oxygenase 1
Gene Aliases:
BCDO, BCDO1, BCMO, BCMO1, BCO
Chromosome Location:
Chr.16: 81238642 - 81291142 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01015946_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: BCO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_017429.2 NP_059125.2 7 - 8 1320 95
XM_017023286.1 XP_016878775.1 7 - 8 1369 95
XM_017023287.1 XP_016878776.1 7 - 8 1369 95
XM_017023288.1 XP_016878777.1 7 - 8 1369 95
XM_017023289.1 XP_016878778.1 3 - 4 434 95
GenBank mRNA AF294900.1 - 7 - 8 1320 95
AK001592.1 - 7 - 8 1320 95
AK296011.1 - 6 - 7 1192 95
BC126210.1 - 7 - 8 1222 8991926 95
BC126212.1 - 7 - 8 1222 8991928 95

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600008577.hg.1
This assay targets the same location as HTA transcript cluster(s) TC16000637.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 86 and 100 3' Most Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene BCO1

Panther Classification:

Molecular Function -

oxygenase

Gene Ontology Categories:

Function(s) Process(es)

retinoid metabolic process
vitamin A biosynthetic process
retinol metabolic process
retinal metabolic process
oxidation-reduction process
beta-carotene metabolic process
beta-carotene 15,15'-monooxygenase activity
metal ion binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR16009016.hg.1 ENST00000258168.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009016.hg.1 ENST00000425577.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR16009016.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR16009016.hg.1 TC16000637.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay PSR1600164003.hg.1 TC1600008577.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000258168.6 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000563804.5 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 ENST00000564552.1 Ensembl
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq
Affymetrix Human Clariom D Assay TC1600008577.hg.1 NM_017429.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0