This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01015969_m1
See other SLC4A7 GE Assays ›
Gene Symbol:
SLC4A7
Gene Name:
solute carrier family 4 member 7
Gene Aliases:
NBC2, NBC3, NBCN1, SBC2, SLC4A6
Chromosome Location:
Chr.3: 27372721 - 27484420 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01015969_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC4A7
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001321103.1 NP_001308032.1 7 - 8 1445 79
NM_001321104.1 NP_001308033.1 7 - 8 1406 79
NM_001321105.1 NP_001308034.1 7 - 8 1406 79
NM_001321106.1 NP_001308035.1 7 - 8 1394 79
NM_003615.4 NP_003606.3 7 - 8 1195 79
NR_135541.1 7 - 8 1394 79
NR_135544.1 7 - 8 1394 79
XM_006713421.2 XP_006713484.1 7 - 8 1433 79
XM_011534256.2 XP_011532558.1 7 - 8 1706 79
XM_011534258.1 XP_011532560.1 7 - 8 1409 79
XM_011534261.2 XP_011532563.1 7 - 8 1671 79
XM_017007526.1 XP_016863015.1 8 - 9 1330 79
XM_017007528.1 XP_016863017.1 1 - 2 104 79
GenBank mRNA AF047033.1 - 7 - 8 1195 79
EU934246.1 - 7 - 8 1321 79
EU934247.1 - 7 - 8 1333 79
EU934248.1 - 7 - 8 1372 79
EU934250.1 - 7 - 8 1333 79
FJ178574.1 - 7 - 8 1321 79
GU354307.1 - 7 - 8 1321 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0300010561.hg.1
This assay targets the same location as HTA transcript cluster(s) TC03001255.hg.1

Additional Information:

This assay is in the 5'UTR region of the XM_017007528.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Transporter Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene SLC4A7

Panther Classification:

Molecular Function -

secondary carrier transporter

Gene Ontology Categories:

Function(s) Process(es)

inorganic anion transport
bicarbonate transport
sodium ion transmembrane transport
regulation of intracellular pH
auditory receptor cell development
anion transmembrane transport
inorganic anion exchanger activity
anion transmembrane transporter activity
sodium:bicarbonate symporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 TC03001255.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000295736.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000388777.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000425128.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000428386.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000435667.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000437179.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000440156.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000445684.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000446700.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000454389.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 ENST00000455077.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR03021521.hg.1 NM_003615.4 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000295736.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000425128.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000428005.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000428179.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000428386.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000437179.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000437266.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000438530.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000440156.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000445684.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000446700.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000454389.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000455077.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000457377.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000491211.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 NM_001258379.1 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 NM_001258380.1 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 NM_003615.4 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000295736.9 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000425128.6 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000428005.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000428179.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000428386.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000437179.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000437266.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000438530.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000440156.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000445684.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000446700.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000454389.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000455077.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000457377.5 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 ENST00000491211.1 Ensembl
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 NM_001258379.1 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 NM_001258380.1 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 NM_003615.4 Refseq
Affymetrix Human Clariom D Assay PSR0300177066.hg.1 TC0300010561.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0