This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01016245_m1
See other ETNPPL GE Assays ›
Gene Symbol:
ETNPPL
Gene Name:
ethanolamine-phosphate phospho-lyase
Gene Aliases:
AGXT2L1
Chromosome Location:
Chr.4: 108742040 - 108763180 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01016245_m1
Size
Availability Made To Order
Catalog # 4426961
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: ETNPPL
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NR_027475.1 1 - 2 237 71
GenBank mRNA DC311077.1 - 1 - 2 212 71
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Transcript Location Amplicon Length
GenBankmRNA DB472042.1 - 1 - 2 158 Chr. 4 - 108759769 - 108763000 [-] on Build GRCh38 71
GenBankmRNA AV647220.1 - 1 - 2 84 Chr. 4 - 108754639 - 108762926 [-] on Build GRCh38 71

Adjacent Gene Details

Gene name Gene Symbol Alias Distance Relative Direction Location Chromsome
Upstream oligosaccharyltransferase complex non-catalytic subunit OSTC DC2 -91.9kb Anti-sense Chr. 4 - 108650585 - 108667822 on NCBI Build GRCh38
Downstream collagen type XXV alpha 1 COL25A1 AMY,
CFEOM5,
CLAC,
CLAC-P,
CLACP
45.7kb Sense Chr. 4 - 108808725 - 109302737 on NCBI Build GRCh38

Back To Top

More Information


Set Membership:

Amplicon length between 71 and 85 Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

phosphatidylethanolamine biosynthetic process
transaminase activity
pyridoxal phosphate binding
identical protein binding
ethanolamine-phosphate phospho-lyase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-cjlz8:80/100.66.74.50:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0