This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01016426_m1
See other RBM10 GE Assays ›
Gene Symbol:
RBM10
Gene Name:
RNA binding motif protein 10
Gene Aliases:
DXS8237E, GPATC9, GPATCH9, S1-1, TARPS, ZRANB5
Chromosome Location:
Chr.X: 47145196 - 47186815 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01016426_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RBM10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001204467.1 NP_001191396.1 4 - 5 828 69
NM_001204468.1 NP_001191397.1 4 - 5 859 69
NM_005676.4 NP_005667.2 4 - 5 828 69
XM_005272677.4 XP_005272734.1 4 - 5 881 69
XM_017029884.1 XP_016885373.1 4 - 5 822 69
XM_017029885.1 XP_016885374.1 4 - 5 597 69
GenBank mRNA AK000962.1 - 5 - 6 773 69
AK225175.1 - 4 - 5 787 69
AL137421.1 - 5 - 6 717 69
BC000681.2 - 1 - 2 53 69
BC004181.2 - 4 - 5 781 2961094 69
BC008733.2 - 4 - 5 813 3163064 69
BC024153.2 - 3 - 4 443 3845258 69
BX537969.1 - 4 - 5 810 69
D50912.1 - 4 - 5 679 69
KU178412.1 - 3 - 4 435 69

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AL137421.1, XM_017029884.1, XM_017029885.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene RBM10

Gene Ontology Categories:

Function(s) Process(es)

regulation of alternative mRNA splicing, via spliceosome
mRNA processing
biological_process
RNA splicing
regulation of apoptotic process
nucleotide binding
protein binding
zinc ion binding
protein complex binding
identical protein binding
poly(A) RNA binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mt7zw:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0