Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
ENST00000359156.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
ENST00000396620.3 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
ENST00000432237.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
ENST00000541972.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
NM_004244.5 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
NM_203416.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR12015460.hg.1 |
TC12001162.hg.1 |
HTA TC |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000359156.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000396620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000432237.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000537044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000537626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000541972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
NM_004244.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
NM_203416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000359156.8 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000396620.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000432237.2 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000537044.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000537626.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
ENST00000541972.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
NM_004244.5 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
NM_203416.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1200202516.hg.1 |
TC1200012749.hg.1 |
Clariom D TC |