This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01017637_g1
See other UXT GE Assays ›
Gene Symbol:
UXT
Gene Name:
ubiquitously expressed prefoldin like chaperone
Gene Aliases:
ART-27, STAP1
Chromosome Location:
Chr.X: 47651792 - 47659180 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01017637_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: UXT
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_004182.3 NP_004173.1 4 - 5 326 68
NM_153477.2 NP_705582.1 3 - 4 504 68
NR_045560.1 4 - 5 326 68
GenBank mRNA AF083241.1 - 4 - 5 292 68
AF083242.1 - 3 - 4 442 68
AF092737.1 - 4 - 5 307 68
AK223320.1 - 4 - 5 280 68
AK312072.1 - 4 - 5 292 68
BC000720.1 - 3 - 4 261 3505415 68
BC008890.1 - 3 - 4 261 3959862 68
EL949569.1 - 4 - 5 304 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0X00009582.hg.1
This assay targets the same location as HTA transcript cluster(s) TC0X001000.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 3' Most Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene UXT

Panther Classification:

Molecular Function -

general transcription factor

Gene Ontology Categories:

Function(s) Process(es)

negative regulation of transcription from RNA polymerase II promoter
microtubule cytoskeleton organization
transcription, DNA-templated
mitochondrion transport along microtubule
centrosome organization
RNA polymerase II transcription corepressor activity
chromatin binding
protein binding
microtubule binding
beta-tubulin binding
actin filament binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 TC0X001000.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 ENST00000333119.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 ENST00000485641.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013290.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR0X013288.hg.1 TC0X001000.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR0X013288.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165306.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165306.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165306.hg.1 TC0X00009582.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay TC0X00009582.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000333119.7 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000335890.2 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000376964.3 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000460840.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 ENST00000485641.5 Ensembl
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NM_004182.3 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NM_153477.2 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NR_045559.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 NR_045560.1 Refseq
Affymetrix Human Clariom D Assay PSR0X00165308.hg.1 TC0X00009582.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-zpsr4:80/100.66.78.116:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0