This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01018102_g1
See other TMEM175 GE Assays ›
Gene Symbol:
TMEM175
Gene Name:
transmembrane protein 175
Gene Aliases:
hTMEM175
Chromosome Location:
Chr.4: 932387 - 958656 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01018102_g1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM175
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297423.1 NP_001284352.1 5 - 6 989 65
NM_001297424.1 NP_001284353.1 3 - 4 305 65
NM_001297425.1 NP_001284354.1 3 - 4 302 65
NM_032326.3 NP_115702.1 5 - 6 528 65
XM_005272301.2 XP_005272358.1 5 - 6 503 65
XM_005272303.4 XP_005272360.1 6 - 7 685 65
XM_005272307.4 XP_005272364.1 5 - 6 454 65
XM_017008701.1 XP_016864190.1 6 - 7 605 65
XM_017008702.1 XP_016864191.1 3 - 4 252 65
XM_017008703.1 XP_016864192.1 6 - 7 1047 65
XM_017008704.1 XP_016864193.1 5 - 6 944 65
GenBank mRNA AK090848.1 - 5 - 6 919 65
AK309132.1 - 5 - 6 454 65
BC005158.2 - 5 - 6 441 3505819 65
BI819231.1 - 3 - 4 249 65
CB992640.1 - 3 - 4 270 65

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC04000014.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene TMEM175

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of lysosomal lumen pH
potassium ion transmembrane transport
phagosome-lysosome fusion
potassium ion leak channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 TC04000014.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 ENST00000264771.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 ENST00000359768.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 ENST00000504180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 ENST00000508204.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 ENST00000515740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000346.hg.1 NM_032326.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 TC04000014.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 ENST00000264771.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 ENST00000359768.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 ENST00000504180.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 ENST00000508204.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 ENST00000515740.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR04000345.hg.1 NM_032326.3 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0