This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01018109_m1
See other TMEM175 GE Assays ›
Gene Symbol:
TMEM175
Gene Name:
transmembrane protein 175
Gene Aliases:
hTMEM175
Chromosome Location:
Chr.4: 932387 - 958656 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01018109_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: TMEM175
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001297426.1 NP_001284355.1 7 - 8 800 68
NM_001297427.1 NP_001284356.1 6 - 7 1056 68
NM_001297428.1 NP_001284357.1 4 - 5 372 68
XM_005272304.1 XP_005272361.1 6 - 7 525 68
XM_017008705.1 XP_016864194.1 3 - 4 228 68
GenBank mRNA AL834199.1 - 7 - 8 670 68
CF242841.1 - 4 - 5 275 68

Back To Top

More Information


Additional Information:

This assay is in the 5'UTR region of the AL834199.1, NM_001297426.1, NM_001297427.1, NM_001297428.1, XM_005272304.1, XM_017008705.1 transcripts.

Set Membership:

Amplicon length less than or equal to 70 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

ion channel

Gene Ontology Categories:

Function(s) Process(es)

regulation of lysosomal lumen pH
potassium ion transmembrane transport
phagosome-lysosome fusion
potassium ion leak channel activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0