| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15014680.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15014680.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15014680.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR15014680.hg.1 |
TC15001549.hg.1 |
HTA TC |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
TC1500009736.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
NM_000942.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
TC1500009736.hg.1 |
Clariom D TC |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
ENST00000300026.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
ENST00000558492.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
ENST00000561048.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR1500171908.hg.1 |
NM_000942.4 |
Refseq |