This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01018972_g1
See other PIEZO1 GE Assays ›
Gene Symbol:
PIEZO1
Gene Name:
piezo type mechanosensitive ion channel component 1
Gene Aliases:
DHS, FAM38A, LMPH3, Mib
Chromosome Location:
Chr.16: 88715338 - 88785220 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01018972_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: PIEZO1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001142864.3 NP_001136336.2 35 - 36 5030 68
GenBank mRNA AB161230.1 - 24 - 25 3481 68
BC150271.1 - 24 - 25 3318 9007243 68
D87071.1 - 24 - 25 3318 68
KC602455.1 - 35 - 36 4774 68

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1600011228.hg.1

Set Membership:

Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

cation transport
positive regulation of integrin activation
positive regulation of cell-cell adhesion mediated by integrin
cation transmembrane transport
cation channel activity
mechanically-gated ion channel activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Clariom D Assay PSR1600187397.hg.1 ENST00000301015.13 Ensembl
Affymetrix Human Clariom D Assay PSR1600187397.hg.1 NM_001142864.3 Refseq
Affymetrix Human Clariom D Assay PSR1600187397.hg.1 ENST00000301015.13 Ensembl
Affymetrix Human Clariom D Assay PSR1600187397.hg.1 NM_001142864.3 Refseq
Affymetrix Human Clariom D Assay PSR1600187397.hg.1 TC1600011228.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-rpjph:80/100.66.75.107:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0