This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01019200_m1
See other CHD1L GE Assays ›
Gene Symbol:
CHD1L
Gene Name:
chromodomain helicase DNA binding protein 1 like
Gene Aliases:
ALC1, CHDL
Chromosome Location:
Chr.1: 147172804 - 147295766 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01019200_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CHD1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001256336.1 NP_001243265.1 11 - 12 1230 125
NM_001256337.1 NP_001243266.1 7 - 8 807 125
NM_001256338.1 NP_001243267.1 6 - 7 725 125
NM_004284.4 NP_004275.4 12 - 13 1337 125
NM_024568.2 NP_078844.2 12 - 13 1309 125
XM_006711639.3 XP_006711702.1 11 - 12 1205 125
XM_017002858.1 XP_016858347.1 13 - 14 2065 125
XM_017002859.1 XP_016858348.1 12 - 13 1192 125
XM_017002860.1 XP_016858349.1 12 - 13 1278 125
XM_017002861.1 XP_016858350.1 11 - 12 1211 125
XM_017002863.1 XP_016858352.1 10 - 11 1026 125
XM_017002864.1 XP_016858353.1 11 - 12 1204 125
GenBank mRNA AF537213.1 - 12 - 13 1294 125
AK001342.1 - 12 - 13 1294 125
AK027631.1 - 12 - 13 1266 125
AK293157.1 - 7 - 8 742 125
AK297135.1 - 6 - 7 663 125
AK307400.1 - 12 - 13 1294 125
BC001171.1 - 12 - 13 1271 125
BC043501.1 - 12 - 13 1255 125
BC077717.1 - 6 - 7 662 6573666 125
CB153921.1 - 1 - 2 124 125
CN255647.1 - 1 - 2 93 125
EF560738.1 - 12 - 13 1305 125
KJ534809.1 - 12 - 13 1274 125

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0100018284.hg
This assay targets the same location as HTA transcript cluster(s) TC01001112.hg

Additional Information:

This assay is in the 5'UTR region of the XM_017002861.1,XM_017002863.1,XM_017002864.1 transcripts.

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CHD1L

Gene Ontology Categories:

Function(s) Process(es)

nucleotide-excision repair, DNA duplex unwinding
DNA repair
nucleotide-excision repair, preincision complex stabilization
nucleotide-excision repair, preincision complex assembly
nucleotide-excision repair, DNA incision, 3'-to lesion
nucleotide-excision repair, DNA incision, 5'-to lesion
chromatin remodeling
cellular response to DNA damage stimulus
nucleotide-excision repair, DNA incision
global genome nucleotide-excision repair
nucleotide binding
nucleic acid binding
ATP-dependent DNA helicase activity
protein binding
ATP binding
ATPase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2xklz:80/100.66.76.139:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0