This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01019209_m1
See other SLC7A10 GE Assays ›
Gene Symbol:
SLC7A10
Gene Name:
solute carrier family 7 member 10
Gene Aliases:
ASC1, HASC-1, asc-1
Chromosome Location:
Chr.19: 33208664 - 33225850 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01019209_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SLC7A10
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011527120.1 XP_011525422.1 1 - 2 277 130
GenBank mRNA AK307527.1 - 1 - 2 240 130

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Panther Classification:

Molecular Function -

transporter

Gene Ontology Categories:

Function(s) Process(es)

amino acid transmembrane transport
amino acid transport
neutral amino acid transport
L-serine transport
D-alanine transport
D-serine transport
leukocyte migration
neutral amino acid transmembrane transporter activity
L-serine transmembrane transporter activity
antiporter activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000253188.8 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000587064.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590036.5 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000590490.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 ENST00000592596.1 Ensembl
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq
Affymetrix Human Clariom D Assay TC1900010379.hg.1 NM_019849.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-7c59b9b899-mnfm6:80/100.66.78.37:80.
git-commit: 8ef7cf686e3c81d28b13492708ce56d167b2f995
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.49.0-Offline