This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01021429_m1
See other CD163 GE Assays ›
Gene Symbol:
CD163
Gene Name:
CD163 molecule
Gene Aliases:
M130, MM130, SCARI1
Chromosome Location:
Chr.12: 7470816 - 7503818 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01021429_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: CD163
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_203416.3 NP_981961.2 15 - 16 3474 122
XM_005253528.4 XP_005253585.1 14 - 15 3563 122
GenBank mRNA AK226017.1 - 15 - 16 3424 122
DQ058615.1 - 15 - 16 3346 122
Z22968.1 - 15 - 16 3432 122
Z22971.1 - 14 - 15 3531 122

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC12001162.hg.1

Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene CD163

Panther Classification:

Molecular Function -

serine protease

Gene Ontology Categories:

Function(s) Process(es)

receptor-mediated endocytosis
acute-phase response
scavenger receptor activity
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 ENST00000359156.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 ENST00000396620.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 ENST00000432237.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 ENST00000541972.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 NM_004244.5 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 NM_203416.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR12015457.hg.1 TC12001162.hg.1 HTA TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-blue-67f85b6989-9hvhj:80/100.66.74.192:80.
git-commit: a6f50562c839d11f1e5940093fdcbf4cc66353c1
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.37.1-Offline