This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01021756_g1
See other HSD11B1L GE Assays ›
Gene Symbol:
HSD11B1L
Gene Name:
hydroxysteroid 11-beta dehydrogenase 1 like
Gene Aliases:
11-DH3, 11-beta-HSD3, HSD3, SCDR10, SCDR10B, SDR26C2
Chromosome Location:
Chr.19: 5680765 - 5688523 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01021756_g1
Size
Availability Made To Order
Catalog # 4351372
Price 4,430.00
Your Price
Online offer:
4430.0
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: HSD11B1L
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001267868.1 NP_001254797.1 5 - 6 975 60
NM_198533.2 NP_940935.1 4 - 5 837 60
NM_198706.2 NP_941995.1 4 - 5 837 60
NM_198707.2 NP_941996.1 3 - 4 774 60
GenBank mRNA AK308206.1 - 5 - 6 715 60
AY268352.1 - 4 - 5 538 60
AY268353.1 - 3 - 4 498 60
AY313894.1 - 4 - 5 566 60

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene HSD11B1L

Panther Classification:

Molecular Function -

dehydrogenase

Gene Ontology Categories:

Function(s) Process(es)

oxidation-reduction process
oxidoreductase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-vh84l:80/100.66.79.51:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0