This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01021872_m1
See other SKP2 GE Assays ›
Gene Symbol:
SKP2
Gene Name:
S-phase kinase-associated protein 2, E3 ubiquitin protein ligase
Gene Aliases:
FBL1, FBXL1, FLB1, p45
Chromosome Location:
Chr.5: 36151989 - 36192792 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01021872_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: SKP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq XM_011514082.2 XP_011512384.1 9 - 10 1335 118

Back To Top

More Information


Set Membership:

Amplicon length greater than or equal to 101 3' Most Homo sapiens Probe spans exons

Gene Ontology Categories:

Function(s) Process(es)

G1/S transition of mitotic cell cycle
G2/M transition of mitotic cell cycle
protein polyubiquitination
cell proliferation
anaphase-promoting complex-dependent catabolic process
positive regulation of intracellular estrogen receptor signaling pathway
regulation of apoptotic process
positive regulation of smooth muscle cell proliferation
regulation of cell cycle
cellular response to cell-matrix adhesion
positive regulation of protein polyubiquitination
ubiquitin-protein transferase activity
protein binding
identical protein binding
ubiquitin protein ligase activity

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-l4hk6:80/100.66.76.55:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0