This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01022071_m1
See other AIMP2 GE Assays ›
Gene Symbol:
AIMP2
Gene Name:
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases:
JTV-1, JTV1, P38
Chromosome Location:
Chr.7: 6009241 - 6023834 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01022071_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIMP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006303.3 NP_006294.2 1 - 2 248 79
GenBank mRNA AK313037.1 - 1 - 2 254 79
BC002853.1 - 1 - 2 208 3634481 79
BC010156.1 - 1 - 2 208 3619946 79
BC013630.1 - 1 - 2 216 3446074 79
U24169.1 - 1 - 2 248 79

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC0700006634.hg.1
This assay targets the same location as HTA transcript cluster(s) TC07000059.hg.1

Set Membership:

Ambion siRNA available Amplicon length between 71 and 85 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AIMP2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
apoptotic process
negative regulation of cell proliferation
positive regulation of protein ubiquitination
Type II pneumocyte differentiation
positive regulation of neuron death
protein binding

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR07000993.hg.1 TC07000059.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07000993.hg.1 ENST00000223029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000993.hg.1 ENST00000400479.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000993.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07000985.hg.1 TC07000059.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07000985.hg.1 ENST00000223029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000985.hg.1 ENST00000400479.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000985.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07000989.hg.1 TC07000059.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07000989.hg.1 ENST00000223029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000989.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000989.hg.1 ENST00000400479.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000989.hg.1 NM_006303.3 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR07000990.hg.1 TC07000059.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 PSR07000990.hg.1 ENST00000223029.3 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000990.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR07000990.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145736.hg.1 TC0700006634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay TC0700006634.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 ENST00000395236.2 Ensembl
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145738.hg.1 TC0700006634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145732.hg.1 TC0700006634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145735.hg.1 TC0700006634.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 ENST00000223029.7 Ensembl
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 ENST00000400479.6 Ensembl
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 ENST00000415999.1 Ensembl
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 NM_006303.3 Refseq
Affymetrix Human Clariom D Assay PSR0700145741.hg.1 TC0700006634.hg.1 Clariom D TC

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-nwsk8:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0