This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01022073_m1
See other AIMP2 GE Assays ›
Gene Symbol:
AIMP2
Gene Name:
aminoacyl tRNA synthetase complex interacting multifunctional protein 2
Gene Aliases:
JTV-1, JTV1, P38
Chromosome Location:
Chr.7: 6009241 - 6023834 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01022073_m1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: AIMP2
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_001326606.1 NP_001313535.1 3 - 4 569 105
NM_001326607.1 NP_001313536.1 2 - 3 482 105
NM_001326609.1 NP_001313538.1 4 - 5 775 105
NM_001326611.1 NP_001313540.1 3 - 4 488 105
NM_006303.3 NP_006294.2 3 - 4 689 105
XM_005249847.3 XP_005249904.1 3 - 4 903 105
XM_005249848.2 XP_005249905.1 3 - 4 579 105
XM_005249850.3 XP_005249907.1 3 - 4 500 105
XM_006715772.2 XP_006715835.1 3 - 4 474 105
XM_011515519.1 XP_011513821.1 3 - 4 504 105
GenBank mRNA AK313037.1 - 3 - 4 695 105
BC002853.1 - 3 - 4 649 3634481 105
BC010156.1 - 3 - 4 649 3619946 105
BC013630.1 - 3 - 4 657 3446074 105
CX867709.1 - 3 - 4 541 105
U24169.1 - 3 - 4 689 105

Back To Top

More Information


Set Membership:

Ambion siRNA available Amplicon length greater than or equal to 101 Mammalian Gene Collection Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene AIMP2

Panther Classification:

Molecular Function -

scaffold/adaptor protein

Gene Ontology Categories:

Function(s) Process(es)

tRNA aminoacylation for protein translation
apoptotic process
negative regulation of cell proliferation
positive regulation of protein ubiquitination
Type II pneumocyte differentiation
positive regulation of neuron death
protein binding

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-2k58s:80/100.66.76.192:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0