This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01022076_m1
See other RAC1 GE Assays ›
Gene Symbol:
RAC1
Gene Name:
ras-related C3 botulinum toxin substrate 1 (rho family, small GTP binding protein Rac1)
Gene Aliases:
MIG5, Rac-1, TC-25, p21-Rac1
Chromosome Location:
Chr.7: 6374495 - 6403967 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01022076_m1
Size
Availability Made To Order
Catalog # 4351372
Price
Your Price
Online offer:
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: RAC1
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
GenBank mRNA BQ689587.1 - 1 - 2 225 63

Back To Top

More Information


Set Membership:

Amplicon length less than or equal to 70 Homo sapiens 1700 Probe spans exons

MicroArray Probes:

Affymetrix ABI

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Transcriptome Array 2.0 TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000348035.8 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000356142.4 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000473564.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000488373.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000495499.1 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 ENST00000497741.5 Ensembl
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_006908.4 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Affymetrix Human Clariom D Assay TC0700006648.hg.1 NM_018890.3 Refseq
Array Name Probe IDs Targets Target Type
ABI Human Genome Survey Microarray Version 2.0 106774 - -

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-lzcpb:80/100.66.79.31:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0