This product has been added to your favorites list. Go to My Favorites

Please enter the information below and press OK to send your cart to Core Services for purchase.

    

System Message

OKCancel
LOADING ...
  • Home
  • › Search Tool
  • › Search Results
  • › Hs01023064_g1
See other DAGLA GE Assays ›
Gene Symbol:
DAGLA
Gene Name:
diacylglycerol lipase alpha
Gene Aliases:
C11orf11, DAGL(ALPHA), DAGLALPHA, NSDDR
Chromosome Location:
Chr.11: 61680184 - 61747002 on Build GRCh38
Species:
Human
Species Specific ID (Flybase ID):
-
Assay ID Hs01023064_g1
Size
Availability Made To Order
Catalog # 4351372
Price 435.00
Your Price
Online offer: 408.65
408.65
Check your price ›
  • Genomic Map
  • Assay Details
  • More Information

Genomic Map

LOADING...
×
Back To Top

Assay Details

Gene Symbol: DAGLA
Interrogated Sequence Translated Protein Exon Boundary Assay Location IMAGE Clone ID Amplicon Length
RefSeq NM_006133.2 NP_006124.1 10 - 11 1244 62
XM_017018238.1 XP_016873727.1 11 - 12 1571 62
XM_017018239.1 XP_016873728.1 11 - 12 1476 62
XM_017018240.1 XP_016873729.1 11 - 12 1571 62
GenBank mRNA AB014559.2 - 10 - 11 1239 62
AY275377.1 - 9 - 10 1128 62
BC150176.1 - 10 - 11 1244 40148003 62
BC150195.1 - 10 - 11 1244 40148734 62
BC152453.1 - 10 - 11 1239 8862586 62

Back To Top

More Information


Updated Mapping Information

This assay targets the same location as Clariom D transcript cluster(s) TC1100007818.hg.1
This assay targets the same location as HTA transcript cluster(s) TC11000538.hg.1

Set Membership:

Ambion siRNA available Amplicon length less than or equal to 70 Mammalian Gene Collection Detect Genomic DNA Homo sapiens Probe spans exons

Matching Ambion siRNAs:

Click here to find siRNAs for Gene DAGLA

Panther Classification:

Molecular Function -

hydrolase lipase metabolite interconversion enzyme

Gene Ontology Categories:

Function(s) Process(es)

neuroblast proliferation
arachidonic acid metabolic process
neurotransmitter biosynthetic process
diacylglycerol catabolic process
endocannabinoid signaling pathway
metal ion binding
acylglycerol lipase activity

MicroArray Probes:

Affymetrix

Array Name Probe IDs Targets Target Type
Affymetrix Human Transcriptome Array 2.0 PSR11006500.hg.1 ENST00000257215.5 Ensembl
Affymetrix Human Transcriptome Array 2.0 PSR11006500.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 PSR11006500.hg.1 TC11000538.hg.1 HTA TC
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Transcriptome Array 2.0 TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay PSR1100155573.hg.1 TC1100007818.hg.1 Clariom D TC
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000257215.9 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 ENST00000540717.1 Ensembl
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq
Affymetrix Human Clariom D Assay TC1100007818.hg.1 NM_006133.2 Refseq

Back To Top

Related Products

  • TaqMan® Fast Advanced Master Mix
  • SuperScript® VILO™ cDNA Synthesis Kit

Your items have has been added!


Host server : magellan-search-green-659f68c6f4-mvcqg:80/100.66.75.199:80.
git-commit: 208ebf2ce40f07c29af7b8d1bec64c518c8c0cf8
git-url: https://github.com/thermofisher/magellan-search
git-branch: release/2.43.0-2026.01.05-1.0